missing translation for 'onlineSavingsMsg'
Learn More

Chloroquine Diphosphate Salt, MP Biomedicals

£57.35 - £97.00

Chemical Identifiers

CAS 50-63-5
Molecular Formula C18H32ClN3O8P2
Molecular Weight (g/mol) 515.86
MDL Number MFCD00069852
InChI Key QKICWELGRMTQCR-UHFFFAOYNA-N
Synonym chloroquine diphosphate, chloroquine phosphate, aralen phosphate, chingamin phosphate, chloroquine bis phosphate, chingaminum, alermine, h-stadur, aralen diphosphate, miniquine
PubChem CID 64927
SMILES OP(O)(O)=O.OP(O)(O)=O.CCN(CC)CCCC(C)NC1=C2C=CC(Cl)=CC2=NC=C1
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11416331
View Documents
MP Biomedicals
0219391910
10 g
£57.35
10g
Please to purchase this item. Need a web account? Register with us today!
11426331
View Documents
MP Biomedicals
0219391925
25 g
£59.40
25g
Please to purchase this item. Need a web account? Register with us today!
11436331
View Documents
MP Biomedicals
0219391950
50 g
£97.00
50g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

50-63-5
515.86
QKICWELGRMTQCR-UHFFFAOYNA-N
64927
C18H32ClN3O8P2
MFCD00069852
chloroquine diphosphate, chloroquine phosphate, aralen phosphate, chingamin phosphate, chloroquine bis phosphate, chingaminum, alermine, h-stadur, aralen diphosphate, miniquine
OP(O)(O)=O.OP(O)(O)=O.CCN(CC)CCCC(C)NC1=C2C=CC(Cl)=CC2=NC=C1

Specifications

50-63-5
MFCD00069852
QKICWELGRMTQCR-UHFFFAOYNA-N
7-chloro-N-[5-(diethylamino)pentan-2-yl]quinolin-4-amine; bis(phosphoric acid)
64927
Chloroquine Diphosphate Salt
C18H32ClN3O8P2
chloroquine diphosphate, chloroquine phosphate, aralen phosphate, chingamin phosphate, chloroquine bis phosphate, chingaminum, alermine, h-stadur, aralen diphosphate, miniquine
OP(O)(O)=O.OP(O)(O)=O.CCN(CC)CCCC(C)NC1=C2C=CC(Cl)=CC2=NC=C1
515.86
515.9
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Chloroquine Diphosphate Salt

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4

Hazard Statement
  • H302-Harmful if swallowed.

Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : Chloroquinone Phosphate

SDS
Documents

Documents

Product Certifications