Learn More
Chlorosulfonic acid, 97%, Thermo Scientific Chemicals
£23.92 - £103.52
Chemical Identifiers
CAS | 7790-94-5 |
---|---|
InChI Key | XTHPWXDJESJLNJ-UHFFFAOYSA-N |
Synonym | chlorosulfonic acid, chlorosulfuric acid, sulfuric chlorohydrin, chlorosulphuric acid, chlorosulphonic acid, monochlorosulfuric acid, unii-2o9axl1tj4, sulfonic acid, monochloride, chloridosulfuric acid, hsdb 909 |
PubChem CID | 24638 |
IUPAC Name | sulfurochloridic acid |
SMILES | OS(=O)(=O)Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10319170
|
Thermo Scientific Acros
304491000 |
100 mL | Glass bottle |
£23.92
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10512851
|
Thermo Scientific Acros
304495000 |
500 mL | Glass bottle |
£31.56
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10053740
|
Thermo Scientific Acros
304490010 |
1 L | Glass bottle |
£49.35
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10397853
|
Thermo Scientific Acros
304490025 |
2.5 L | Glass bottle |
£103.52
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7790-94-5 | |
chlorosulfonic acid, chlorosulfuric acid, sulfuric chlorohydrin, chlorosulphuric acid, chlorosulphonic acid, monochlorosulfuric acid, unii-2o9axl1tj4, sulfonic acid, monochloride, chloridosulfuric acid, hsdb 909 | |
sulfurochloridic acid |
XTHPWXDJESJLNJ-UHFFFAOYSA-N | |
24638 | |
OS(=O)(=O)Cl |
Specifications
7790-94-5 | |
151°C to 152°C | |
97% | |
ClSO3H | |
1754 | |
15, 2168 | |
Solubility in water: reacts with water forming sulfuric and hydrochlori. Other solubilities: de,decomposes in acids,decomposes in alcohol,insoluble in cs2 | |
OS(=O)(=O)Cl | |
116.52 | |
116.52 | |
Chlorosulfonic acid |
− 80.0°C | |
1.7500g/mL | |
HClO3S | |
Oily Liquid, Fuming In Air | |
01,140; 02,70; 06,121 | |
chlorosulfonic acid, chlorosulfuric acid, sulfuric chlorohydrin, chlorosulphuric acid, chlorosulphonic acid, monochlorosulfuric acid, unii-2o9axl1tj4, sulfonic acid, monochloride, chloridosulfuric acid, hsdb 909 | |
XTHPWXDJESJLNJ-UHFFFAOYSA-N | |
sulfurochloridic acid | |
24638 | |
97% |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes severe skin burns and eye damage.
Reacts violently with water.
GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 232-234-6
- Chlorosulfonic acid
Signal Word
- Danger
Hazard Category
- Skin corrosion/irritation Category 1 A
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H335-May cause respiratory irritation.
- H314-Causes severe skin burns and eye damage.
- EUH014-Reacts violently with water.
Precautionary Statement
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
RUO – Research Use Only