Learn More
Chlorothiazide, 98%, Thermo Scientific Chemicals
£26.60 - £147.00
Chemical Identifiers
CAS | 58-94-6 |
---|---|
Molecular Formula | C7H6ClN3O4S2 |
Molecular Weight (g/mol) | 295.71 |
MDL Number | MFCD00058576 |
InChI Key | JBMKAUGHUNFTOL-UHFFFAOYSA-N |
Synonym | chlorothiazide, diuril, chlorthiazide, chlorothiazid, chlotride, chlortiazid, thiazide, flumen, chlorosal, chlorurit |
PubChem CID | 2720 |
ChEBI | CHEBI:3640 |
IUPAC Name | 6-chloro-1,1-dioxo-4H-1$l^{6},2,4-benzothiadiazine-7-sulfonamide |
SMILES | NS(=O)(=O)C1=CC2=C(NC=NS2(=O)=O)C=C1Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15783447
|
Thermo Scientific Acros
461740050 |
5 g |
£26.60
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15793447
|
Thermo Scientific Acros
461740250 |
25 g |
£46.80
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15703457
|
Thermo Scientific Acros
461741000 |
100 g |
£147.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
58-94-6 | |
295.71 | |
JBMKAUGHUNFTOL-UHFFFAOYSA-N | |
2720 | |
6-chloro-1,1-dioxo-4H-1$l^{6},2,4-benzothiadiazine-7-sulfonamide |
C7H6ClN3O4S2 | |
MFCD00058576 | |
chlorothiazide, diuril, chlorthiazide, chlorothiazid, chlotride, chlortiazid, thiazide, flumen, chlorosal, chlorurit | |
CHEBI:3640 | |
NS(=O)(=O)C1=CC2=C(NC=NS2(=O)=O)C=C1Cl |
Specifications
58-94-6 | |
97.5% min. (HPLC) | |
MFCD00058576 | |
Solubility in water: very slightly soluble. Other solubilities: freely soluble in dmso, dmf, slightly soluble in methanol, pyridine, practically insoluble in diethyl ether, benzene, and chloroform | |
NS(=O)(=O)C1=CC2=C(NC=NS2(=O)=O)C=C1Cl | |
295.71 | |
CHEBI:3640 | |
98% |
348.0°C | |
C7H6ClN3O4S2 | |
chlorothiazide, diuril, chlorthiazide, chlorothiazid, chlotride, chlortiazid, thiazide, flumen, chlorosal, chlorurit | |
JBMKAUGHUNFTOL-UHFFFAOYSA-N | |
6-chloro-1,1-dioxo-4H-1$l^{6},2,4-benzothiadiazine-7-sulfonamide | |
2720 | |
295.72 | |
Chlorothiazide |
Safety and Handling
GHS H Statement
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
GHS P Statement
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice/attention.
In case of inadequate ventilation wear respiratory protecti
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 200-404-9
RUO – Research Use Only