missing translation for 'onlineSavingsMsg'
Learn More
Chlorotrimethylsilane, 98%, AcroSeal™ Product Code.: 10433575

Chlorotrimethylsilane, 98%, AcroSeal™

Product Code. 10433575
100 mL, AcroSeal™ Glass bottle
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 

Product Code. 10433575

Brand: Thermo Scientific Acros 426431000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 

CAS: 75-77-4 | C3H9ClSi | 108.64 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Silylation reagent
TRUSTED_SUSTAINABILITY

Specifications

CAS 75-77-4
Melting Point -58.0°C
Color Colorless
Density 0.8500g/mL
Boiling Point 57.0°C
Flash Point −28°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging AcroSeal™ Glass bottle
Molecular Formula C3H9ClSi
Linear Formula (CH3)3SiCl
Refractive Index 1.3870 to 1.3890
MDL Number MFCD00000502
Quantity 100 mL
Merck Index 15, 2180
Specific Gravity 0.85
Synonym trimethylchlorosilane, trimethylsilyl chloride, silane, chlorotrimethyl, trimethyl chlorosilane, monochlorotrimethylsilicon, tmcs, chloro trimethyl silane, silane, trimethylchloro, silicane, chlorotrimethyl, tmscl
Solubility Information Solubility in water: reacts. Other solubilities: care must be taken with ketones,soluble in benzene,ether and perchloroethylene
InChI Key IJOOHPMOJXWVHK-UHFFFAOYSA-N
SMILES C[Si](C)(C)Cl
IUPAC Name chloro(trimethyl)silane
Molecular Weight (g/mol) 108.64
PubChem CID 6397
ChEBI CHEBI:85069
Viscosity 0.34 mPa.s (25°C)
Formula Weight 108.64
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Chlorotrimethylsilane
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0467351 Certificate TypeCertificate of Analysis Date31/10/2024 Catalog Number426431000
Lot NumberA0457745 Certificate TypeCertificate of Analysis Date09/11/2023 Catalog Number426431000
Lot NumberA0445411 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number426431000
Lot NumberA0403087 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number426431000
Lot NumberA0439406 Certificate TypeCertificate of Analysis Date09/08/2022 Catalog Number426431000
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Chlorotrimethylsilane
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H301-Toxic if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH071-Corrosive to the respiratory tract.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Chlorotrimethylsilane, 98%, AcroSeal™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.