missing translation for 'onlineSavingsMsg'
Learn More
Chlorotrimethylsilane, 98%, Thermo Scientific Chemicals Product Code.: 10214520

Chlorotrimethylsilane, 98%, Thermo Scientific Chemicals

Product Code. 10214520
25 mL, Glass bottle
Click to view available options
Quantity:
25 mL
100 mL
250 mL
1 L
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10214520

Brand: Thermo Scientific Acros 110120250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Silylation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-77-4
Molecular Formula C3H9ClSi
Molecular Weight (g/mol) 108.64
MDL Number MFCD00000502
InChI Key IJOOHPMOJXWVHK-UHFFFAOYSA-N
Synonym trimethylchlorosilane, trimethylsilyl chloride, silane, chlorotrimethyl, trimethyl chlorosilane, monochlorotrimethylsilicon, tmcs, chloro trimethyl silane, silane, trimethylchloro, silicane, chlorotrimethyl, tmsclShow MoreShow Less
PubChem CID 6397
ChEBI CHEBI:85069
IUPAC Name chloro(trimethyl)silane
SMILES C[Si](C)(C)Cl

Specifications

Melting Point -58.0°C
Color Colorless
Density 0.8500g/mL
Boiling Point 57.0°C
Flash Point −28°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)3SiCl
Refractive Index 1.3870 to 1.3890
Quantity 25 mL
Beilstein 04,IV,4007
Fieser 01,1232; 02,435; 03,310; 04,537; 05,709; 06,626; 07,66; 08,107; 09,112; 10,96; 11,127; 12,279; 13,165; 14,88; 15,14; 16,23; 17,79
Merck Index 15,218
Specific Gravity 0.85
Solubility Information Solubility in water: reacts. Other solubilities: care must be taken with ketones,soluble in benzene,ether and perchloroethylene
Viscosity 0.34 mPa.s (25°C)
Formula Weight 108.64
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Chlorotrimethylsilane
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0462601 Certificate TypeCertificate of Analysis Date07/05/2024 Catalog Number110120010,110121000,110122500,110120250
Lot NumberA0450049 Certificate TypeCertificate of Analysis Date05/02/2024 Catalog Number110120010,110121000,110122500,110120250
Lot NumberA0459555 Certificate TypeCertificate of Analysis Date24/01/2024 Catalog Number110120010,110121000,110120025,110122500,110120250
Lot NumberA0456596 Certificate TypeCertificate of Analysis Date26/09/2023 Catalog Number110120010,110121000,110120025,110122500,110120250
Lot NumberA0400255 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number110120010,110121000,110120025,110122500,110120250
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Chlorotrimethylsilane
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 A
  • Acute toxicity Category 4
  • Acute toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
  • H332-Harmful if inhaled.
  • H301-Toxic if swallowed.
  • H312-Harmful in contact with skin.
  • EUH014-Reacts violently with water.
  • EUH071-Corrosive to the respiratory tract.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P402+P404-Store in a dry place. Store in a closed container.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Chlorotrimethylsilane, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.