missing translation for 'onlineSavingsMsg'
Learn More

Chlorotriphenylsilane, 96%, Thermo Scientific Chemicals

Product Code. 11423900
Click to view available options
Quantity:
5 g
25 g
50 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chlorotriphenylsilane is used as a silylating agent. It is also used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chlorotriphenylsilane is used as a silylating agent. It is also used as a pharmaceutical intermediate.

Solubility
Reacts with water.

Notes
Keep container tightly sealed. Store under dry inert gas. It is sensitive to moisture. Incompatible with bases and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76-86-8
Molecular Formula C18H15ClSi
Molecular Weight (g/mol) 294.85
MDL Number MFCD00000496
InChI Key MNKYQPOFRKPUAE-UHFFFAOYSA-N
Synonym triphenylsilyl chloride, triphenylchlorosilane, silane, chlorotriphenyl, triphenylsilicon chloride, chloro triphenyl silane, triphenylsilylchloride, chloro-tri phenyl silane, benzene, 1,1',1-chlorosilylidyne tris, clsiph3, ph3sicl
PubChem CID 6458
IUPAC Name chloro(triphenyl)silane
SMILES Cl[Si](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Boiling Point 188°C to 189°C (3mmHg)
Melting Point 91°C to 96°C
Quantity 50 g
UN Number UN3261
Beilstein 1820487
Flash Point >200°C (392°F)
Solubility Information Reacts with water.
Formula Weight 294.86
Percent Purity 96%
Odor Pungent
Sensitivity Moisture sensitive
Chemical Name or Material Chlorotriphenylsilane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'