missing translation for 'onlineSavingsMsg'
Learn More

chlorotripyrrolidinophosphonium hexafluorophosphate, 98%, Thermo Scientific™

Product Code. p-4258698 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11986671

Brand: Thermo Scientific Acros 438640050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Coupling reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 133894-48-1
Molecular Formula C12H24ClN3P·F6P
Molecular Weight (g/mol) 421.73
InChI Key BSCYRXJVGSZNKX-UHFFFAOYSA-N
Synonym pyclop, chlorotripyrrolidinophosphonium hexafluorophosphate, chlorotripyrrolidinophosphonium hexa-fluorophosphate, chlorotri 1-pyrrolidinyl phosphonium hexafluorophosphate, 98+%, pyciop, ref dupl: pyclop, chlorotripyrrolidino, acmc-1c01w, ksc177a1n, chlorotripyrrolidinophosphoniumhexafluorophosphat
PubChem CID 16211753
IUPAC Name chloro(tripyrrolidin-1-yl)phosphanium;hexafluorophosphate
SMILES C1CCN(C1)[P+](N2CCCC2)(N3CCCC3)Cl.F[P-](F)(F)(F)(F)F

Specifications

Melting Point 145.0°C to 148.0°C
Color White to Yellow
Physical Form Crystalline Powder
Quantity 5 g
Assay Percent Range 97.5% min. (HPLC)
Formula Weight 421.73
Percent Purity 98%
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material Chlorotripyrrolidinophosphonium hexafluorophosphate
compliance-icons
Product Identifier
  • Chlorotripyrrolidinophosphonium hexafluorophosphate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.