missing translation for 'onlineSavingsMsg'
Learn More

Chlorotris(triphenylphosphine)cobalt(I), 97%, Thermo Scientific Chemicals

Product Code. 15441618
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

A stoichiometric reducing agent that is used in the radical dimerization of halogenated organic molecules.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A stoichiometric reducing agent that is used in the radical dimerization of halogenated organic molecules.

Solubility
Insoluble in water.

Notes
Air & Moisture Sensitive.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26305-75-9
Molecular Formula C54H45ClCoP3
Molecular Weight (g/mol) 881.26
MDL Number MFCD00015864
InChI Key RHNBOEDTKLKUEB-UHFFFAOYSA-M
Synonym chlorotris triphenylphosphine cobalt i, cobalt i tris triphenylphosphine chloride
PubChem CID 71431130
IUPAC Name cobalt;triphenylphosphane;chloride
SMILES [Cl-].[Co+].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 135°C to 139°C
Quantity 5 g
Sensitivity Air and moisture sensitive
Solubility Information Insoluble in water.
Formula Weight 881.26
Percent Purity 97%
Chemical Name or Material Chlorotris(triphenylphosphine)cobalt(I)

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.