missing translation for 'onlineSavingsMsg'
Learn More

Chlortetracycline hydrochloride, specified according to the requirements of Ph.Eur., Thermo Scientific Chemicals

Chlortetracycline hydrochloride, CAS # 64-72-2, is a broad-spectrum antibiotic of the tetracycline family.

£28.54 - £226.13

Chemical Identifiers

CAS 64-72-2
Molecular Formula C22H23ClN2O8·HCl
Molecular Weight (g/mol) 515.33
MDL Number MFCD00082440
InChI Key QYAPHLRPFNSDNH-CIVPRPTRSA-N
Synonym chlortetracycline hydrochloride, 4-epi-chlortetracycline hydrochloride, 2-amino hydroxy methylidene-7-chloro-4-dimethylamino-6,10,11,12a-tetrahydroxy-6-methyl-4,4a,5,5a-tetrahydrotetracene-1,3,12-trione hydrochloride, 2-azanyl oxidanyl methylidene-7-chloranyl-4-dimethylamino-6-methyl-6,10,11,12a-tetrakis oxidanyl-4,4a,5,5a-tetrahydrotetracene-1,3,12-trione hydrochloride
PubChem CID 66577600
IUPAC Name (4S,4aR,5aS,6S,12aR)-7-chloro-4-(dimethylamino)-1,6,10,11,12a-pentahydroxy-6-methyl-3,12-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carboxamide;hydrochloride
SMILES CC1(C2CC3C(C(=O)C(=C(C3(C(=O)C2=C(C4=C(C=CC(=C41)Cl)O)O)O)O)C(=O)N)N(C)C)O.Cl
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10882041
View Documents
Thermo Scientific Acros
430960050
5 g Glass bottle
£28.54
5g
Please to purchase this item. Need a web account? Register with us today!
10785652
View Documents
Thermo Scientific Acros
430960250
25 g Glass bottle
£79.60
25g
Please to purchase this item. Need a web account? Register with us today!
10220130
View Documents Promotion Details
Thermo Scientific Acros
430961000
100 g Glass bottle
£226.13
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General features
• Chlortetracycline hydrochloride is a broad-spectrum antibiotic of the tetracycline family
• Receives its name from a 7-chloro substitution on one of the rings of its parent structure, tetracycline
• A bacteriostatic that inhibits the 30S ribosomal subunit and prevents peptide formation
• Can work as an antiprotozoal and antifungal


Applications
• Chlortetracycline is suitable for use in biochemistry, microbiology, in vivo research, and cell culture
• Intended for research use only

Specifications

Chemical Identifiers

64-72-2
515.33
QYAPHLRPFNSDNH-CIVPRPTRSA-N
66577600
CC1(C2CC3C(C(=O)C(=C(C3(C(=O)C2=C(C4=C(C=CC(=C41)Cl)O)O)O)O)C(=O)N)N(C)C)O.Cl
C22H23ClN2O8·HCl
MFCD00082440
chlortetracycline hydrochloride, 4-epi-chlortetracycline hydrochloride, 2-amino hydroxy methylidene-7-chloro-4-dimethylamino-6,10,11,12a-tetrahydroxy-6-methyl-4,4a,5,5a-tetrahydrotetracene-1,3,12-trione hydrochloride, 2-azanyl oxidanyl methylidene-7-chloranyl-4-dimethylamino-6-methyl-6,10,11,12a-tetrakis oxidanyl-4,4a,5,5a-tetrahydrotetracene-1,3,12-trione hydrochloride
(4S,4aR,5aS,6S,12aR)-7-chloro-4-(dimethylamino)-1,6,10,11,12a-pentahydroxy-6-methyl-3,12-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carboxamide;hydrochloride

Specifications

64-72-2
220.0°C
2.3 to 3.3 (1% H2O, CO2 free)
C22H23ClN2O8·HCl
15,2198
− 242.50
Solubility in water: 8.6g/L (28°C). Other solubilities: practically insoluble in acetone,ether,,chloroform,dioxane and propylene glycol,17.4g/L methanol (28°C),1.7g/L ethanol (28°C),soluble in solutions of alkali hydroxides and,carbonat
CC1(C2CC3C(C(=O)C(=C(C3(C(=O)C2=C(C4=C(C=CC(=C41)Cl)O)O)O)O)C(=O)N)N(C)C)O.Cl
515.33
515.33
Powder
0.40 max. (460nm)
Yellow
Authentic
MFCD00082440
− 242.50 (20.00°C c=0.5,water)
chlortetracycline hydrochloride, 4-epi-chlortetracycline hydrochloride, 2-amino hydroxy methylidene-7-chloro-4-dimethylamino-6,10,11,12a-tetrahydroxy-6-methyl-4,4a,5,5a-tetrahydrotetracene-1,3,12-trione hydrochloride, 2-azanyl oxidanyl methylidene-7-chloranyl-4-dimethylamino-6-methyl-6,10,11,12a-tetrakis oxidanyl-4,4a,5,5a-tetrahydrotetracene-1,3,12-trione hydrochloride
QYAPHLRPFNSDNH-CIVPRPTRSA-N
(4S,4aR,5aS,6S,12aR)-7-chloro-4-(dimethylamino)-1,6,10,11,12a-pentahydroxy-6-methyl-3,12-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carboxamide;hydrochloride
66577600
94.5 to 102.0% (sum of the contents of chlortetra cycline hydrochloride and tetracycline hydrochloride, dry substance), ≥89.5% (chlortetracycline hydrochloride, dry substance), 8% max. (tetracycline hydrochloride, dry substance)
Chlortetracycline hydrochloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 200-591-7

Product Identifier
  • Chlortetracycline hydrochloride

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.