missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ CHO CD EfficientFeed™ B AGT™ Nutrient Supplement
CHO CD EfficientFeed™ B AGT™ Nutrient Supplement
£1534.00 - £11170.00
Specifications
Content And Storage | Dry format medium Store at 2 to 8°C, Store Dark and Dry |
---|---|
Cell Line | CHO |
Classification | Animal Origin-free, Chemically-defined, Protein-free, Serum-free |
Culture Type | Suspension Cell Culture |
Form | AGT |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11380892
|
Gibco™
A1245605 |
10 L |
£1534.00
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11370892
|
Gibco™
A1245601 |
100 L |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Gibco™ CHO CD EfficientFeed™ B AGT™ nutrient supplement is a single part dry format product for fed batch addition to the culture of multiple cell lines. CHO CD EfficientFeed™ B AGT™ (Advanced Granulation Technology™) nutrient supplement is an animal origin-free, chemically defined formulation that contains no proteins, hydrolysates, or components of incompletely defined composition.The AGT™ format is designed for large scale manufacturing and ease of use. This product is also available in liquid format for small scale use and experimentation.
Which EfficientFeed™ Nutrient Supplement to Choose?
Consider the CHO CD Efficient Feed™ Kit when:
Consider CHO CD EfficientFeed™ A supplement when:
Consider CHO CD EfficientFeed™ B supplement when:
Consider CD EfficientFeed™ C AGT™ supplement when:
Increased Titers
CHO CD EfficientFeed™ supplements have been shown to increase titers 2-5 fold versus batch culture, and are an excellent starting point for process development (Figures 1a & 1b). Furthermore, they outperform competitive alternatives, including hydrolysates (Figures 2, 3a, & 3b).
Simple Scale-Up and Cost Savings
The ability to scale up in a dry format provides ease of handling and storage in manufacturing and other bioprocess settings. With AGT™ (Advanced Granulation Technology™) dry format, it is possible to store and use the product even more easily and safely than traditional dry format products. CHO CD EfficientFeed™ B AGT™ nutrient supplement is offered in 10 L and 100 L packaging that allows scale-up from the 1 L catalog liquid as a seamless process, as well as allow for cost savings when purchasing production level volumes.
Comparable Performance to Liquid Supplement
Equally important as the convenience and simplicity of AGT™ (Advanced Granulation Technology™) dry format is the comparable performance to the liquid CHO CD EfficientFeed™ B (Figure 4). Both forms have performed similarly in our testing, and the AGT™ version has been used successfully as a prototype in many processes by multiple companies.
AGT™ Format Ease of Use
CHO CD EfficientFeed™ B AGT™ nutrient supplement delivers dry format scale-up convenience as well as simple reconstitution via water addition and a single pH adjustment step (see Product Profile Sheet below for complete instructions).
For research or further manufacturing use. Not for use in diagnostic procedures.
Specifications
Dry format medium Store at 2 to 8°C, Store Dark and Dry |
|
Animal Origin-free, Chemically-defined, Protein-free, Serum-free | |
AGT | |
Serum-Free | |
Sodium Pyruvate, Sodium Bicarbonate | |
Low | |
Chemically Defined Nutrient Feeding Supplement | |
Solubility |
CHO | |
Suspension Cell Culture | |
Nutrient Supplement | |
Non-sterile | |
No Glutamine, No Phenol Red, No Insulin | |
Cell Culture Grade, Qualified for Further Manufacturing, Research Grade | |
Wet Ice |
For Research Use or Further Manufacturing. Not for use in diagnostic procedures.