missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ CHO DG44 Cells (cGMP banked) and Media Kit
Click to view available options
Quantity:
1 kit
Unit Size:
Each
Description
The CHO DG44 Cells (cGMP banked) and Media Kit is optimized for the growth of dihydrofolate reductase deficient (DHFR-) Chinese Hamster Ovary (CHO) cells and expression of recombinant proteins in suspension culture. The kit provides:
GIBCO™ CD DG44 Medium—a chemically defined, protein-free medium specifically designed to enable optimal growth and expression of DG44 cells in suspension culture
CHO DG44 cells (cGMP banked) —Parental CHO DG44 cells have been produced, banked and tested to meet cGMP quality standards. They are pre-adapted to CD DG44 Medium and selected for superior cell growth and transfection efficiencies.
In addition, the CHO DG44 Cells (cGMP banked) and Media Kit includes L-glutamine and Pluronic™ F-68 for optimal growth and protein expression of DG44 cells in a protein-free, chemically defined environment.
GIBCO™ CD DG44 Medium—a chemically defined, protein-free medium specifically designed to enable optimal growth and expression of DG44 cells in suspension culture
CHO DG44 cells (cGMP banked) —Parental CHO DG44 cells have been produced, banked and tested to meet cGMP quality standards. They are pre-adapted to CD DG44 Medium and selected for superior cell growth and transfection efficiencies.
In addition, the CHO DG44 Cells (cGMP banked) and Media Kit includes L-glutamine and Pluronic™ F-68 for optimal growth and protein expression of DG44 cells in a protein-free, chemically defined environment.
Specifications
Specifications
| Classification | Animal Origin-Free, Chemically Defined, Protein-Free, Serum-Free |
| Content And Storage | CHO DG44 Cells (cGMP banked) and Media Kit (SKU A1100001) contains: • CD DG44 Medium 1000 mL- 12610-010, store in the dark at 2-8°C • CHO DG44 Cells (cGMP banked) 1 vial (> 1x10^7 cells) - A1097101 (see limited label license), store in liquid nitrogen • PLURONIC F-68, 10% (100X) 100 mL - 24040-032, store at room temperature • L-glutamine-200mM (100X) 100 mL - 25030-081 or 25030-024 (CE marked version for Europe only), store at -20°C |
| Cell Line | CHO DG44 |
| Culture Type | Suspension Cell Culture |
| Species | Hamster |
| Media Recommendation | CD DG44 Medium |
| No. of Cells | 1 x 107 |
| Product Line | Gibco |
| Quantity | 1 kit |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction