missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ CHO-S-SFM II, with hypoxanthine and thymidine
CHO-S-SFM II, with hypoxanthine and thymidine
£80.00 - £139.00
Specifications
Content And Storage | Store in refrigerator (2–8°C) and protect from light. |
---|---|
Cell Line | CHO |
Classification | Low Protein, Serum-free |
Concentration | 1 X |
Culture Type | Suspension Cell Culture |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11580416
|
Gibco™
12052114 |
500 mL |
£80.00
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10503483
|
Gibco™
12052098 |
1 L |
£139.00
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CHO-S-SFM II, is a complete, serum-free, low-protein (less than 100μ/mL) medium optimized for the growth and maintenance of Chinese hamster ovary (CHO) cells in suspension culture for production of recombinant proteins. It is supplemented with hypoxanthine and thymidine (the non-supplemented equivalent is also available). This ready-to-use medium is suitable for the suspension culturing of CHO and other cells in batch, continuous, and perfusion culture systems. A CHO-S-SFM II medium Drug Master File (DMF) has been submitted to the FDA. Permission to cross-reference the Master File may be obtained by contacting Technical Services or your local Sales Representative.This medium contains components of animal origin. If growth factors or hydrolysates, found in competitor media, are to be avoided, consider CD CHO, CD OptiCHO™ or CD FortiCHO™ media.
When to use CHO-S-SFM II Medium
When to Consider Another Gibco™ Medium
Consider CD FortiCHO™ medium when:
Consider CD OptiCHO™ medium when:
Consider CD CHO medium when:
Consider CD DG44 medium when:
Design Your Own Media Specific to Your Needs
Whether it’s a bench-level experiment or large scale biomanufacturing process, we offer specialized formats, packaging, and quality levels specific to your needs. With Gibco™ cGMP Media Custom Services you can put the manufacturing power and years of technical experience of our R&D and Bioproduction Application Specialists to work for you.
Specifications
Store in refrigerator (2–8°C) and protect from light. | |
Low Protein, Serum-free | |
Suspension Cell Culture | |
Serum Free Medium (SFM) | |
Low | |
Sustainable packaging | |
Gibco™ |
CHO | |
1 X | |
Liquid | |
Serum-free | |
Bottle(s) | |
No antibiotics | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.