missing translation for 'onlineSavingsMsg'
Learn More

Berkshire™ Choice™ Cleanroom Face Masks

Three-layer cleanroom face masks provide filtration efficiency while providing long wearing comfort.

£953.00

Specifications

Product Type Cleanroom Face Mask
Color White
Fastening Type Earloops
Material Polypropylene
Sterility Non-sterile
View More Specs

Products 1
Product Code Brand Packaging Type Price Quantity & Availability  
Product Code Brand Packaging Type Price Quantity & Availability  
18849410
missing translation for 'viewDocuments'
Berkshire™
CHFM18040B
Bulk Packaged
£953.00
Pack of 2000
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • 3-Layer cleanroom masks
  • Knitted nylon earloops
  • Filter medium: meltblown polypropylene provides particle and bacterial filtration
  • Fully enclosed nosepiece is adjustable
  • Available in bulk or individually wrapped packaging
Specifications

Specifications

Cleanroom Face Mask
Earloops
Non-sterile
ASTM F2299
Class 10 (ISO 4)
Single-use
Choice™
White
Polypropylene
Pleated
EN 14683
3-ply
99.7% (PFE), 99.7% (BFE)
Videos
SDS
Documents

Documents

Certificates