missing translation for 'onlineSavingsMsg'
Learn More

Cholesteryl benzoate, 98%, Thermo Scientific Chemicals

£45.12 - £133.67

Chemical Identifiers

CAS 604-32-0
Molecular Formula C34H50O2
Molecular Weight (g/mol) 490.77
MDL Number MFCD00003635
InChI Key UVZUFUGNHDDLRQ-UHFFFAOYNA-N
Synonym cholesteryl benzoate, cholesterol benzoate, 3b-benzoyloxy cholest-5-ene, unii-n09h13shlb, n09h13shlb, cholest-5-ene-3-beta-yl benzoate, cholest-5-en-3-ol 3.beta.-, benzoate, 5-cholesten-3.beta.-ol benzoate, cholest-5-en-3-ol 3beta-, benzoate
PubChem CID 2723613
SMILES CC(C)CCCC(C)C1CCC2C3CC=C4CC(CCC4(C)C3CCC12C)OC(=O)C1=CC=CC=C1
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10132280
View Documents
Thermo Scientific Acros
110200250
25 g Glass bottle
£45.12
25g
Please to purchase this item. Need a web account? Register with us today!
10045923
View Documents
Thermo Scientific Acros
110201000
100 g Glass bottle
£133.67
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

604-32-0
490.77
UVZUFUGNHDDLRQ-UHFFFAOYNA-N
2723613
C34H50O2
MFCD00003635
cholesteryl benzoate, cholesterol benzoate, 3b-benzoyloxy cholest-5-ene, unii-n09h13shlb, n09h13shlb, cholest-5-ene-3-beta-yl benzoate, cholest-5-en-3-ol 3.beta.-, benzoate, 5-cholesten-3.beta.-ol benzoate, cholest-5-en-3-ol 3beta-, benzoate
CC(C)CCCC(C)C1CCC2C3CC=C4CC(CCC4(C)C3CCC12C)OC(=O)C1=CC=CC=C1

Specifications

604-32-0
White
97.5% min. (HPLC)
MFCD00003635
-15
Solubility in water: insoluble. Other solubilities: soluble in chloroform,methylene chloride,,benzene,toluene,diethyl ether
CC(C)CCCC(C)C1CCC2C3CC=C4CC(CCC4(C)C3CCC12C)OC(=O)C1=CC=CC=C1
490.77
490.76
Crystalline Powder
149°C to 150°C
Authentic
C34H50O2
10, 2181
cholesteryl benzoate, cholesterol benzoate, 3b-benzoyloxy cholest-5-ene, unii-n09h13shlb, n09h13shlb, cholest-5-ene-3-beta-yl benzoate, cholest-5-en-3-ol 3.beta.-, benzoate, 5-cholesten-3.beta.-ol benzoate, cholest-5-en-3-ol 3beta-, benzoate
UVZUFUGNHDDLRQ-UHFFFAOYNA-N
9a,11a-dimethyl-1-(6-methylheptan-2-yl)-1H,2H,3H,3aH,3bH,4H,6H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-yl benzoate
2723613
98%
Cholesteryl benzoate
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

missing translation for 'einecsNumber' : 210-064-3

Product Identifier
  • Cholesteryl benzoate

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only