missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Choline chloride, 98+%, Thermo Scientific Chemicals
16.15 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Use promo code "21743" to get your promotional price.
Click to view available options
Quantity:
100 g
500 g
2500 g
Description
Choline chloride helps in providing nourishment for pig feed and poultry feed. It acts as a food flavor enhancer. It is used as a precursor of acetylcholine and is a methyl donor in various metabolic processes and in lipid metabolism.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Choline chloride helps in providing nourishment for pig feed and poultry feed. It acts as a food flavor enhancer. It is used as a precursor of acetylcholine and is a methyl donor in various metabolic processes and in lipid metabolism.
Solubility
Soluble in water, ethanol, methanol and alcohol.
Notes
Incompatible with strong oxidizing agents, strong bases, strong acids, alkalies and alloys.
Choline chloride helps in providing nourishment for pig feed and poultry feed. It acts as a food flavor enhancer. It is used as a precursor of acetylcholine and is a methyl donor in various metabolic processes and in lipid metabolism.
Solubility
Soluble in water, ethanol, methanol and alcohol.
Notes
Incompatible with strong oxidizing agents, strong bases, strong acids, alkalies and alloys.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | ∽305°C (decomposition) |
Assay Percent Range | ≥98% |
Quantity | 100 g |
Beilstein | 3563126 |
Sensitivity | Hygroscopic |
Merck Index | 14,2206 |
Solubility Information | Soluble in water,ethanol,methanol and alcohol. |
Formula Weight | 139.63 |
Percent Purity | ≥98% |
Physical Form | Solid |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'