Learn More
Choline Chloride, 99%, Thermo Scientific Chemicals
£19.00 - £273.00
Chemical Identifiers
CAS | 67-48-1 |
---|---|
Molecular Formula | C5H14ClNO |
Molecular Weight (g/mol) | 139.62 |
MDL Number | MFCD00011721 |
InChI Key | SGMZJAMFUVOLNK-UHFFFAOYSA-M |
Synonym | choline chloride, hepacholine, biocolina, biocoline, lipotril, paresan, luridin chloride, choline hydrochloride, cholinium chloride, bilineurin chloride |
PubChem CID | 6209 |
ChEBI | CHEBI:133341 |
IUPAC Name | 2-hydroxyethyl(trimethyl)azanium;chloride |
SMILES | [Cl-].C[N+](C)(C)CCO |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10011973
|
Thermo Scientific Acros
110290500 |
50 g | Plastic bottle |
£19.00
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10570181
|
Thermo Scientific Acros
110295000 |
500 g | Plastic bottle |
£44.80
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10715561
|
Thermo Scientific Acros
110290010 |
1 kg | Plastic bottle |
£74.20
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10703924
|
Thermo Scientific Acros
110290050 |
5 kg | Plastic drum |
£273.00
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
67-48-1 | |
139.62 | |
SGMZJAMFUVOLNK-UHFFFAOYSA-M | |
6209 | |
2-hydroxyethyl(trimethyl)azanium;chloride |
C5H14ClNO | |
MFCD00011721 | |
choline chloride, hepacholine, biocolina, biocoline, lipotril, paresan, luridin chloride, choline hydrochloride, cholinium chloride, bilineurin chloride | |
CHEBI:133341 | |
[Cl-].C[N+](C)(C)CCO |
Specifications
67-48-1 | |
White | |
98.5 to 101.5% (on dry substance) (Argentometry) | |
HOCH2CH2N(CH3)3Cl | |
04, II, 722 | |
choline chloride, hepacholine, biocolina, biocoline, lipotril, paresan, luridin chloride, choline hydrochloride, cholinium chloride, bilineurin chloride | |
SGMZJAMFUVOLNK-UHFFFAOYSA-M | |
2-hydroxyethyl(trimethyl)azanium;chloride | |
6209 | |
139.62 | |
Adhering Crystals |
298.0°C to 304.0°C | |
Authentic | |
C5H14ClNO | |
MFCD00011721 | |
15, 2211 | |
Solubility in water: soluble. Other solubilities: soluble in ethanol | |
[Cl-].C[N+](C)(C)CCO | |
139.62 | |
CHEBI:133341 | |
99% | |
Choline chloride |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
missing translation for 'einecsNumber' : 200-655-4
RUO – Research Use Only