missing translation for 'onlineSavingsMsg'
Learn More

CHPG, Tocris Bioscience™

Product Code. 30001178
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

mGlu5 selective agonist

A selective mGlu5 metabotropic glutamate receptor agonist, completely inactive at mGlu1a receptors expressed in CHO cells. Active in vivo.

Sodium Salt also available.

Chemical Identifiers

CAS 170846-74-9
Molecular Formula C8H8ClNO3
Molecular Weight (g/mol) 201.606
InChI Key UNIDAFCQFPGYJJ-UHFFFAOYSA-N
Synonym chpg, 2-chloro-5-hydroxyphenylglycine, rs-2-chloro-5-hydroxyphenylglycine, 2-amino-2-2-chloro-5-hydroxyphenyl acetic acid, chlorohydroxyphenylglycine, d08nqw, r,s-chpg, chpg hplc , white solid, r-2-2-chloro-5-hydroxyphenyl glycine, amino 2-chloro-5-hydroxyphenyl acetic acid
PubChem CID 3645780
IUPAC Name 2-amino-2-(2-chloro-5-hydroxyphenyl)acetic acid
SMILES C1=CC(=C(C=C1O)C(C(=O)O)N)Cl

Specifications

Formula Weight 201.61
Percent Purity >99%
Quantity 10 mg
Chemical Name or Material CHPG
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'