missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHRNA10 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55291-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody is specific to CHRNA10.
Ionotropic receptor with a probable role in the modulation of auditory stimuli. Agonist binding may induce an extensive change in conformation that affects all subunits and leads to opening of an ion-conducting channel across the plasma membrane. The channel is permeable to a range of divalent cations including calcium, the influx of which may activate a potassium current which hyperpolarizes the cell membrane. In the ear, this may lead to a reduction in basilar membrane motion, altering the activity of auditory nerve fibers and reducing the range of dynamic hearing. This may protect against acoustic trauma.Specifications
CHRNA10 | |
Polyclonal | |
Unconjugated | |
CHRNA10 | |
CHRNA10, NACHR alpha 10, NACHRA10 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
504186, 57053, 64574 | |
-20°C | |
Liquid |
Western Blot | |
0.41 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9GZZ6, Q9JLB5 | |
Chrna10 | |
Peptide | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction