missing translation for 'onlineSavingsMsg'
Learn More

Plier Style Decapper

Use this decapper with 20mm crimp vials. JG Finneran™ Plier Style Decappers allow for safe and easy removal of caps from vials with a 20mm diameter.

Brand:  JG Finneran Associates 9310-20

Product Code. 10157220

  • £221.00 / Each
Estimated Shipment: 19-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Stainless steel Decapper
  • For 20mm crimp vials
Specifications

Specifications

20 mm Plier-style Decapper
20 mm Caps
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Plier Style Decapper >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.