missing translation for 'onlineSavingsMsg'
Learn More

Chroma Gesellschaft™ Crimp Vial Decapper

Sturdy, hand-operated decapper. Chroma Gesellschaft™ Crimp Vial Decapper easily removes sealed 11mm caps.

Brand:  Chroma Gesellschaft™ 9310-11

Product Code. 10085590

  • £127.00 / Each
Estimated Shipment: 10-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Hand-operated
  • Sturdy construction
  • Designed for removing 11mm caps
Specifications

Specifications

11m crimped caps
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers