missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell™ Microflex™ 93-260 Chemical-Resistant Disposable Glove
18.30 GBP valid until 2024-07-31
Use promo code "21674" to get your promotional price.
Use promo code "21674" to get your promotional price.
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
Description
- Three layer design for superior protection against harsh chemicals including acids, solvents and bases
- Thin mil construction provides enhanced tactility and dexterity
- Extra soft material and ergonomic design for outstanding fit, feel and flexibility for longer wear time
- Lower acceptable pinhole rate (0.65 AQL) and extended cuff for reliable protection against hazardous substances
- Silicone free formulation and processing ensure better product protection
Specifications
Certifications/Compliance | ASTM D3577, TYPE II, PPE 89/686, EN 420, 388 & 374 CATEGORY III |
Color | Green, Blue |
Disposable | Single-use |
Hand | Ambidextrous |
Material | Nitrile, Neoprene |
Powdered | Powder Free |
Product Type | Disposable Gloves |
Size | Large |
Thickness (Metric) | 0.198 mm |
Finger Style | Textured |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'