missing translation for 'onlineSavingsMsg'
Learn More

Minigrip™ Resealable Write-on Bags

Label bag contents with ease. Minigrip™ Resealable Write-on Bags provide three white bars specifically designed for writing on with inkpen or marker.

£19.45 - £95.58

Specifications

Description Resealable Bag
Color Clear
For Use With (Application) Ideal for storing stationery and sending samples
Material Polyethylene
Quantity 100 Pack
View More Specs

Products 7
Product Code Brand Width (Metric) Height (Metric) Price Quantity & Availability  
Product Code Brand Width (Metric) Height (Metric) Price Quantity & Availability  
11326924
View Documents
Minigrip™
BAJ-340-011X
75 mm 85 mm
£23.03
Pack of 100
In Stock
to see stock.
Edge
Add to basket
 
10205074
View Documents
Minigrip™
BAJ-340-051C
100 mm 140 mm
£33.48
Pack of 100
In Stock
to see stock.
Edge
Add to basket
 
10001044
View Documents
Minigrip™
BAJ-340-060B
140 mm 140 mm
£36.59
Pack of 100
In Stock
to see stock.
Edge
Add to basket
 
10366984
View Documents
Minigrip™
BAJ-340-091N
190 mm 190 mm
£67.95
Pack of 100
In Stock
to see stock.
Edge
Add to basket
 
10634185
View Documents
Minigrip™
BAJ-340-003N
40 mm 65 mm
£19.45
Pack of 100
In Stock
to see stock.
Edge
Add to basket
 
10758235
View Documents
Minigrip™
BAJ-340-065X
75 mm 190 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
30057395
View Documents
Minigrip™
BAJ-340-065X
75 mm 190 mm
£95.58
Pack of 1000
Estimated Shipment: 06-05-2025
to see stock.
Add to basket
 
Description

Description

  • Polyethylene
  • Resealable
  • Autoclavable
  • Three write-on bars for labelling
Specifications

Specifications

Resealable Bag
Ideal for storing stationery and sending samples
100 Pack
Clear
Polyethylene
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Minigrip™ Resealable Write-on Bags

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.