missing translation for 'onlineSavingsMsg'
Learn More

Azpack™ Steel Spheres for Viscometer

Helps with the measurement of fluid thickness. Azpack™ Steel Spheres for Viscometer are made of carbon chrome steel.

Brand:  Azpack™ 10608922

 View more versions of this product

Product Code. 10608922

  • £17.90 / Pack of 100
Estimated Shipment: 13-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Constructed of carbon chrome steel
  • Round shape
  • Available in 4 sizes
  • For use with viscometer
  • Density: 7.785g/nL
Specifications

Specifications

Spare Steel Balls
Viscometer
Steel
10 to 375 Strokes
1.58mm dia.
10 to 375 Strokes
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers