Learn More
Macherey-Nagel™ Chromabond™ BT 120 SiOH Column
Brand: Macherey-Nagel™ 732965
This item is not returnable.
View return policy
Specifications
255 mm | |
Flash Column | |
Biotage Isolera Systems |