Learn More
Macherey-Nagel™ Chromabond™ Collecting Rack
Collection rack for use with SPE columns
£229.92 - £232.00
Specifications
For Use With (Application) | Evaporation of Eluates |
---|---|
Type | Collecting Rack |
Product Code | Brand | For Use With (Application) | For Use With (Equipment) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Application) | For Use With (Equipment) | Price | Quantity & Availability | ||||
10368501
|
Macherey-Nagel™
730366 |
Evaporation of Eluates | 16 LV Columns |
£232.00
Each |
Estimated Shipment: 02-09-2024
Log in to see stock. |
||||
10114661
|
Macherey-Nagel™
730157 |
Evaporation of Eluates | 12 Columns |
£232.00
Each |
Estimated Shipment: 02-09-2024
Log in to see stock. |
||||
10072921
|
Macherey-Nagel™
730153 |
Evaporation of Eluates | 24 Columns |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
CHROMABOND™ vacuum manifold for simultaneous preparation of up to 12 samples. The manifold consists of a rectangular glass cabinet with a vacuum gauge and a polypropylene lid, that can hold the columns or cartridges. The replaceable valves on the lid allow individual vacuum control for each solid phase extraction column, if required. The cabinet is fitted with a variable rack with exchangeable partitions, that accept a wide variety of vessels like test tubes, measuring flasks, scintillation vials, autosampler vials, plastic vials and many more. The vacuum in the chamber can be adjusted with the control valve and read from the gauge.Specifications
Evaporation of Eluates | |
Collecting Rack |