Learn More
Macherey-Nagel™ Chromabond™ Easy Polypropylene SPE Column
Brand:  Macherey-Nagel™ 730755
 View more versions of this product
Product Code. 10776522
  • £130.00 / Pack of 30
Estimated Shipment: 21-11-2024
to see stock.
Explore more special offers
Add to basket
Specifications
6 mL
200 mg
80 μm
Solid Phase Extraction Column
650 to 700 m2/g
SPE
Solid Phase Extraction
Chromabond Easy
50 μm