Learn More
Macherey-Nagel™ Chromabond™ Florisil Glass SPE Column
Brand: Macherey-Nagel™ 730239G
This item is not returnable.
View return policy
Specifications
Magnesium Silicate | |
SPE | |
150 to 250 μm | |
Solid Phase Extraction Column |
6 mL | |
2 g | |
Chromabond Florisil |