missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Chromabond™ HR-XC Polypropylene SPE Column

Brand:  Macherey-Nagel™ 730953

Code : 50

Additional Details : Weight : 0.20000kg

 View more versions of this product

Product Code. 12881031

  • £153.00 / Pack of 30
Estimated Shipment: 03-09-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

PS/DVB Copolymer
SPE
Solid Phase Extraction
Chromabond HR-XC
Spherical
800 m2/g
3 mL
500 mg
85 μm
Solid Phase Extraction Column
65 to 75 Å
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers