Learn More
Macherey-Nagel™ Chromabond™ Polyethylene Frit
Product Code. 10072871 Shop All Macherey-Nagel Products
Click to view available options
Volume (Metric):
1 mL
3 mL
6 mL
15 mL
30 mL
45 mL
70 mL
150 mL
Unit Size:
Pack of 250
Product Code. 10072871
Brand: Macherey-Nagel™ 730034
Only null left
Add to basket
Add to basket
Assemble SPE (solid-phase extraction) columns selecting from a range of accessories. Chromabond™ Polyethylene Frits for polyethelyne columns come in a varitey of sizes.
Accessory for SPE (solid-phase extraction)
For individual packing of SPE columns with Chromabond™ adsorbents
Specifications
For Use With (Equipment) Polypropylene Columns
Type Frit
Volume (Metric) 30 mL
Material Polyethylene
Quantity 250 Pack