Learn More
Macherey-Nagel™ Chromabond™ Polyethylene Frit
Assemble SPE (solid-phase extraction) columns selecting from a range of accessories. Chromabond™ Polyethylene Frits for polyethelyne columns come in a varitey of sizes.
Brand:  Macherey-Nagel™ 730164
 View more versions of this product
Product Code. 10338311
  • £113.00 / Pack of 250
Estimated Shipment: 05-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Accessory for SPE (solid-phase extraction)  
For individual packing of SPE columns with ChromabondTM adsorbents
Specifications
Specifications
Polypropylene Columns
1 mL
250 Pack
Frit
Polyethylene