Learn More
Macherey-Nagel™ Chromabond™ Tubing Adapter
For use with vacuum manifolds
Brand:  Macherey-Nagel™ 730387
Product Code. 12860771
  • £226.00 / Pack of 4
Estimated Shipment: 13-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
CHROMABOND™ vacuum manifold for simultaneous preparation of up to 12 samples. The manifold consists of a rectangular glass cabinet with a vacuum gauge and a polypropylene lid, that can hold the columns or cartridges. The replaceable valves on the lid allow individual vacuum control for each solid phase extraction column, if required. The cabinet is fitted with a variable rack with exchangeable partitions, that accept a wide variety of vessels like test tubes, measuring flasks, scintillation vials, autosampler vials, plastic vials and many more. The vacuum in the chamber can be adjusted with the control valve and read from the gauge.
Specifications
Specifications
3 and 6 mL Glass Columns, Application of Large Sample Volumes
Tubing Adapter
4 Pack
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Macherey-Nagel™ Chromabond™ Tubing Adapter > For Use With: For 3 and 6mL Glass Columns, For Application of Large Sample Volumes