missing translation for 'onlineSavingsMsg'
Learn More

ChromaCare™ LC-MS Aqueous Rinse, Probe Wash 1, Thermo Scientific™

Ready-to-use blend of water and 2% acetonitrile (v/v) serves as an aqueous wash solution to clean sample probes and needles on LC instruments.

Brand:  Thermo Scientific T001252500

104.00 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 15842544

  • £107.00 / 2.50 L

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Suitable for cleaning sample preparation needle and drain lines on LC instrument after sampling
  • Save time by using off-the-shelf blend — premade for convenience
  • Helps to maintain clean sample lines and reduce sample carryover
  • Packaged in 2.5L amber glass bottle
  • Complementary to T001262500
TRUSTED_SUSTAINABILITY
Specifications

Specifications

75-05-8
Water with 2% Acetonitrile
2.5 L
1.8 to 2.2% (Acetonitrile)
Amber Glass Bottle
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot Number187700 Certificate TypeCertificate of Analysis Date20/10/2023 Catalog NumberT001252500
Lot Number195882 Certificate TypeCertificate of Analysis Date20/10/2023 Catalog NumberT001252500
Lot Number198163 Certificate TypeCertificate of Analysis Date20/10/2023 Catalog NumberT001252500
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
ChromaCare™ LC-MS Aqueous Rinse, Probe Wash 1, Thermo Scientific™ > 2.5L

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.