missing translation for 'onlineSavingsMsg'
Learn More

ChromaCare™ LC-MS Organic Rinse, Probe Wash 2, Thermo Scientific™

Premade and ready-to-use water-acetonitrile blend (60:40) with 0.1% formic acid serves as a wash solution to clean sample and liquid chromatography injection probes. Commonly used as an auto-sampler cleaning solution in LC-MS.

Brand:  Thermo Scientific T001262500

217.00 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 15852544

  • £223.00 / 2.50 L

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Suitable as a cleaning solution to clean injection needle on LC instrument
  • Save time by using off-the-shelf blend -- pre-made for convenience
  • Helps to maintain clean sample lines and reduce sample carryover
  • Commonly used as auto-sampler cleaning solution in LC-MS
  • Packaged in 2.5L amber glass bottle
TRUSTED_SUSTAINABILITY
Specifications

Specifications

7732-18-5, 64-18-6, 75-05-8
Amber Glass Bottle
Liquid
Water with 40% Acetonitrile and 0.1% Formic Acid
2.5 L
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot Number177755 Certificate TypeCertificate of Analysis Date20/10/2023 Catalog NumberT001262500
Lot Number184206 Certificate TypeCertificate of Analysis Date20/10/2023 Catalog NumberT001262500
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
ChromaCare™ LC-MS Organic Rinse, Probe Wash 2, Thermo Scientific™ > 2.5L

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.