missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Chromacol 12mm and 13mm Closures for Screw-Thread Vials
Thermo Scientific™ Chromacol Vial Closures are offered with a selection of liner materials and colors.
Brand: Thermo Scientific™ 12-SCWG
This item is not returnable.
View return policy
Description
Thermo Scientific™ Chromacol Vial Closures are offered with a selection of liner materials and colors.
Specifications
12mm Screw Cap | |
Polypropylene | |
Screw Thread Vials |
Green | |
Chromacol |