Learn More
Thermo Scientific™ Chromacol™ GOLD-Grade Inert Vials and Inserts
Ensure sample security and integrity with inert vials that have the highest recovery rates, an ultra-low pH shift, and lowest alkali content.
£13.55 - £466.00
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
11573760
|
Thermo Scientific™
01-CVG |
0.1mL Sci-Vi Crimp Top Vial - GOLD Grade Glass |
£253.02
Pack of 500 |
In Stock
Log in to see stock. |
|||||
15232670
|
Thermo Scientific™
2-SVWGK |
2mL Wide Opening SureStop Screw Thread Vial with Write-on Patch, Flat Bottom, GOLD Grade Glass, Clear |
£13.55
Pack of 100 |
Estimated Shipment: 19-08-2024
Log in to see stock. |
|||||
11543760
|
Thermo Scientific™
02-CTVG |
0.2mL Sci-Vi Crimp Top Vial - GOLD Grade Glass |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11543680
|
Thermo Scientific™
2-SVG |
2mL Screw Thread Vial, Flat Bottom, GOLD Grade Glass, Clear |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11503680
|
Thermo Scientific™
1.1-STVG |
8-425 Screw Thread 1.1mL Vial - GOLD Grade Glass |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11553760
|
Thermo Scientific™
03-CVG |
0.3mL Crimp Top, Round Bottom Vial, Clear Gold Grade |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
12631505
|
Thermo Scientific™
02-FISVG |
0.2mL Wide Opening Screw Thread Vial, Insert Vial, Fused Insert-GOLD Grade Glass, Clear |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11533760
|
Thermo Scientific™
2-CVG |
2mL Wide Opening Crimp Top Vial, Flat Bottom, GOLD Grade Glass, Clear |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11503760
|
Thermo Scientific™
1.1-CTVG |
11mm Crimp Top 1.1mL Vial, Wide Opening - GOLD Grade Glass |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
12611505
|
Thermo Scientific™
02-FIVG |
0.2mL Crimp Top Vial with Write-on Patch, Insert Vial, Fused Insert-GOLD Grade Glass, Clear |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
12306808
|
Thermo Scientific™
02-FIRVG |
0.2mL Snap Cap Vial, Fused Conical, Fused Insert-GOLD Grade Glass, Clear |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
10570303
|
Thermo Scientific™
02-MTVWG |
0.2mL Insert, Pulled Point, GOLD Grade Glass, Clear |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Maximize your sample performance and have fewer failed assays with Thermo Scientific™ Chromacol™ GOLD-Grade Inert Glass Vials. These vials offer the most state-of-the-art technology in the industry for the analysis of critical low detection limit analytes, and when sample quality, reproducibility, security, and integrity are of upmost importance.
For easier sample handling and less risk of sample loss, the Chromacol GOLD-Grade Vials are the best-in-class choice for all chromatography users performing ultra-trace analysis.
Achieve confidence in results- Made of 1st hydrolytic class, special 33 expansion glass; meeting current USP requirements for pharmacopeias.
- Ultra-low adsorption surface for critical polar compounds, enabling ultra trace-level analysis for strongly adsorbing analytes.
- The high-class standard in Europe and Asia for decades.
- Ultra-low pH shift due to the minimized surface activity.
- Lowest alkaline content for less glass-wall interactions.
- Highest recovery rates, even with critical compounds (e.g., trisubstituted N-atoms, tertiary amines) in trace concentrations.
- Peaks show the same heights and symmetry and feature high recovery and reproducibility.
- Stable analysis and higher reproducibility means increased security and integrity of analytical results.
- Reduced amount of buffer saves money.
- No risk of run-stops during overnight autosampler runs.
- Lot-independent, optimal autosampler fit.
Specifications
Clear | |
Glass |