missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Chromafix™ C18 ec Cartridge

Product Code. p-8012071 Shop All Macherey-Nagel Products
Click to view available options
Bed Weight:
270 mg
530 mg
950 mg
Unit Size:
Pack of 50
This item is not returnable. View return policy

Product Code. 10348071

Brand: Macherey-Nagel™ 731804

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

Base Material Silica
Carbon Load 14%
Bed Weight 270 mg
For Use With (Application) Non-Polar Compounds, Aflatoxins, Amphetamines, Antibiotics, Antiepileptics, Barbiturates, Caffeine, Drugs, Preservatives, Fatty Acids, Nicotine, PAHs, Pesticides, PCBs, Heavy Metals, Vitamins, Very Well Suited for Desalting of Samples
Particle Size 45 μm
Sorbent C18 ec
Type Chromafix C18 ec Cartridge
Endcapped Yes
Pore Size 60 Å
Quantity 50 Pack
Surface Area (Metric) 500 m2/g
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.