Learn More
Macherey-Nagel™ Chromafix™ Dry (Na2SO4) Cartridge
Brand:  Macherey-Nagel™ 731854
Code : 50
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 10639961
  • £170.00 / Pack of 50
Estimated Shipment: 30-08-2024
to see stock.
Add to basket
Specifications
Dry (Na2 SO4)
Removal of Traces of Water From Organic Solutions
50 Pack
2800 mg
Chromafix Dry (Na2 SO4) Cartridge