Learn More
Macherey-Nagel™ Chromafix™ HR-P Cartridge
Resolve a number of components from water or oil. Chromafix™ HR-P Cartridges contain highly porous polystyrene-divinylbenzene adsorbent resin for SPE.
Brand:  Macherey-Nagel™ 731839
 View more versions of this product
Product Code. 10678572
  • £158.00 / Pack of 50
Estimated Shipment: 12-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Features
  • Highly porous polystyrene-divinylbenzene copolymer
  • Specific surface 1200 m2/g
  • Particle size 50–100 μm
  • Very high binding capacity, up to 30 percent of adsorbent weight (for comparison: silica adsorbents about 3 percent)
Recommended application:
  • Aromatic compounds
  • Phenols from water
  • Nitroaromatics from water
  • Pesticides from water
  • PAHs from oil
Specifications
Specifications
200 mg
50 to 100 μm
Chromafix HR-P Cartridge
Polystyrene-divinylbenzene adsorbent resin
Aromatic Compounds, Phenols From Water, Nitroaromatics From Water, Pesticides From Water, PAHs From Oil
HR-P
50 Pack
1200 m2/g