missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ Chromafix™ HR-XA Cartridge
£192.00 - £272.00
Specifications
Base Material | Spherical PS/DVB Copolymer |
---|---|
For Use With (Application) | Acidic Active Ingredients From Strongly Matrix-Contaminated Samples Like, e.g., Urine, Plasma, Serum, Phenolic Acids, Acidic Herbicides |
Particle Size | 45 or 85 μm |
Sorbent | Strong quat. ammonium modified polymeric anion exchanger |
Type | Chromafix HR-XA Cartridge |
Product Code | Brand | Bed Weight | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Bed Weight | Price | Quantity & Availability | |||||
12824371
|
Macherey-Nagel™
731768 |
155 mg |
£192.00
Pack of 50 |
Estimated Shipment: 09-08-2024
Log in to see stock. |
|||||
12804441
|
Macherey-Nagel™
731769 |
240 mg |
£213.00
Pack of 50 |
Estimated Shipment: 09-08-2024
Log in to see stock. |
|||||
12814511
|
Macherey-Nagel™
731770 |
500 mg |
£272.00
Pack of 50 |
Estimated Shipment: 09-08-2024
Log in to see stock. |
|||||
Specifications
Spherical PS/DVB Copolymer | |
45 or 85 μm | |
Chromafix HR-XA Cartridge | |
50 Pack |
Acidic Active Ingredients From Strongly Matrix-Contaminated Samples Like, e.g., Urine, Plasma, Serum, Phenolic Acids, Acidic Herbicides | |
Strong quat. ammonium modified polymeric anion exchanger | |
55 to 65 Å | |
850 m2/g |