missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ Chromafix™ HR-XAW Cartridge
£192.00 - £272.00
Specifications
Base Material | Spherical PS/DVB Copolymer |
---|---|
For Use With (Application) | Perfluorinated Surfactants, Acidic Compounds Like Sulfonates, Active Ingredients From Strongly Matrix-Contaminated Samples Like, e.g., Urine, Plasma, Serum |
Particle Size | 45 or 85 μm |
Sorbent | Weakly basic secondary and tertiary ammonium polymeric anion exchanger |
Type | Chromafix HR-XAW Cartridge |
Product Code | Brand | Bed Weight | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Bed Weight | Price | Quantity & Availability | |||||
12778252
|
Macherey-Nagel™
731772 |
240 mg |
£213.00
Pack of 50 |
Estimated Shipment: 09-08-2024
Log in to see stock. |
|||||
12768252
|
Macherey-Nagel™
731771 |
155 mg |
£192.00
Pack of 50 |
Estimated Shipment: 09-08-2024
Log in to see stock. |
|||||
12788252
|
Macherey-Nagel™
731773 |
500 mg |
£272.00
Pack of 50 |
Estimated Shipment: 09-08-2024
Log in to see stock. |
|||||
Specifications
Spherical PS/DVB Copolymer | |
45 or 85 μm | |
Chromafix HR-XAW Cartridge | |
50 Pack |
Perfluorinated Surfactants, Acidic Compounds Like Sulfonates, Active Ingredients From Strongly Matrix-Contaminated Samples Like, e.g., Urine, Plasma, Serum | |
Weakly basic secondary and tertiary ammonium polymeric anion exchanger | |
55 to 65 Å | |
850 m2/g |