missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ Chromafix™ HR-XC Cartridge
£199.00 - £272.00
Specifications
Base Material | Spherical PS/DVB Copolymer |
---|---|
For Use With (Application) | Basic Active Ingredients From Strongly Matrix-Contaminated Samples Like, e.g., Urine, Plasma, Serum, Fungicides From Food |
Particle Size | 45 or 85 μm |
Sorbent | Benzenesulfonic acid modified polymeric cation exchanger |
Type | Chromafix HR-XC Cartridge |
Product Code | Brand | Bed Weight | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Bed Weight | Price | Quantity & Availability | |||||
12894301
|
Macherey-Nagel™
731757 |
500 mg |
£272.00
Pack of 50 |
Estimated Shipment: 09-08-2024
Log in to see stock. |
|||||
12830961
|
Macherey-Nagel™
731755 |
155 mg |
£199.00
Pack of 50 |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||||
12801041
|
Macherey-Nagel™
731756 |
240 mg |
£213.00
Pack of 50 |
Estimated Shipment: 09-08-2024
Log in to see stock. |
|||||
Specifications
Spherical PS/DVB Copolymer | |
45 or 85 μm | |
Chromafix HR-XC Cartridge | |
50 Pack |
Basic Active Ingredients From Strongly Matrix-Contaminated Samples Like, e.g., Urine, Plasma, Serum, Fungicides From Food | |
Benzenesulfonic acid modified polymeric cation exchanger | |
65 to 75 Å | |
800 m2/g |