Learn More
Macherey-Nagel™ Chromafix™ PS-OH- cartridges
Brand: Macherey-Nagel™ 731868
Additional Details : Weight : 0.01000kg
Specifications
Strong PS/DVB Cation Exchanger, OH- form | |
100 μm | |
100 Å |
Removal or Concentration of Anions From Water Increasing The pH Value in Acidic Samples | |
Chromafix PS-OH- Cartridge | |
50 Pack |