missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chroman-6-carbonyl chloride, 95%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Specifications
Specifications
CAS Min % | 94.5 |
CAS Max % | 100.0 |
Assay Percent Range | 95% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Formula Weight | 196.63 |
Percent Purity | 95% |
Chemical Name or Material | Chroman-6-carbonyl chloride |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Chroman-6-carbonyl chloride, 95%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction