missing translation for 'onlineSavingsMsg'
Learn More
Learn More
chroman-8-carbonyl chloride, Thermo Scientific™
missing translation for 'orderingAttributeHoverText'
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Specifications
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Formula Weight | 196.63 |
Percent Purity | 90% |
Chemical Name or Material | chroman-8-carbonyl chloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'