missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chromate Standard For Ion Chromatography, SPEX CertiPrep™
Click to view available options
Quantity:
125 mL
Packaging:
Triple-leached LDPE bottle
Description
Includes
NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Analyte | Concentration |
---|---|
CrO4-2 | 1000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
CAS | 7789-00-6, 7732-18-5, 7789-00-6 |
Packaging | Triple-leached LDPE bottle |
Quantity | 125 mL |
CAS Max % | 0.1, 99.9 |
Concentration or Composition Notes | Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. |
For Use With (Equipment) | AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation |
Molecular Formula | CrK2O4 |
MDL Number | MFCD00011368 |
InChI Key | XMXNVYPJWBTAHN-UHFFFAOYSA-N |
SMILES | [K+].[K+].[O-][Cr]([O-])(=O)=O |
Show More |
Safety and Handling
Product Identifier
- 1000
- Danger
- Carcinogenicity Category 1B
- Germ cell mutagenicity Category 1B
- H340-May cause genetic defects.
- H350-May cause cancer.
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : potassium chromate
Product intended for laboratory use only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'