Learn More
Lenz Laborglasinstrumente™ Chromatographic Columns with Frit and Beaded Rim
Brand:  Lenz Laborglasinstrumente™ 05445704
 View more versions of this product
Product Code. 10778461
  • £84.85 / Each
Estimated Shipment: 29-04-2025
to see stock.
Explore more special offers
Add to basket
Specifications
Chromatography Column
20 mm
125 mL
Duran Glass
PTFE Stopcock
Chromatographic applications
Chromatography Column
400 mm
20 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Lenz Laborglasinstrumente™ Chromatographic Columns with Frit and Beaded Rim > Column Capacity: 125mL