missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Chromatography Paper, MN 260

Achieve reproducible retention in thin layer chromatography. Macherey-Nagel™ Chromatography Paper, MN 260 are used to achieve chamber saturation in TLC applications. Each pack contains 100 sheets of 75 x 170mm paper.

Brand:  Macherey-Nagel™ 814030

28.20 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 10083071

  • £36.15 / Pack of 100
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

To achieve reproducible retention in thin layer chromatography applications, saturation of the chamber with solvent vapor is advantageous in many cases. To obtain saturation, a TLC developing chamber can be lined with absorptive chromatography paper.
Specifications

Specifications

Chromatography paper
MN 260
75 mm
For Chamber Saturation
170 mm
TLC
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Macherey-Nagel™ Chromatography Paper, MN 260 > Size: 75 x 170mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.