Learn More
Macherey-Nagel™ ChromCart Guard Column System
Brand:  Macherey-Nagel™ 721359
Product Code. 10249361
  • £59.00 / Each
Estimated Shipment: 30-01-2025
to see stock.
Explore more special offers
Add to basket
Specifications
EC Guard Column 8/3 with 2/3mm I.D., 8/4 with 4/4.6mm I.D.
ChromCart Guard Column System