missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pro-Lab Diagnostics™ Nichrome Inoculation Loops
Click to view available options
Diameter (Metric) Inside:
1.59 mm
2.26 mm
2.52 mm
2.91 mm
3.57 mm
5.05 mm
Gauge:
Heavy
Standard
Unit Size:
Pack of 25
Description
An inoculation loop is a simple tool used to gather and transfer microscopic organisms from a culture or broth plate. The small loop removes a consistent amount of inoculum by using surface tension. The inoculation loop is flame-sterilized before and after each use to eliminate cross-contamination between cultures. (The loop must be cooled after sterilization to avoid destroying the organisms.)
- Constructed of economical nickel-chrome in standard or heavy gauge wire
- Available in a range of 6 sizes, 1μL / 1.59mmD to 10μL / 5.05mmD
- Packaged in sets of 25 loops
- Holders with insulated handles sold separately
Specifications
Specifications
| Diameter (Metric) Inside | 3.57 mm |
| Material | Chrome-Nickel |
| Product Type | Inoculating Loop |
| Gauge | Heavy |
| Quantity | 25/Pk. |
| Calibrated | Yes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction