missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chromeazurol S, Honeywell™
Click to view available options
Quantity:
25 g
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
CAS | 1667-99-8 |
Molecular Formula | C23H13Cl2Na3O9S |
MDL Number | MFCD00001615 |
UN Number | NONH for all modes of transport |
Beilstein | 4110655 |
InChI Key | FUIZKNBTOOKONL-UHFFFAOYSA-K |
SMILES | CC1=CC(=CC(=C1[O-])C(=O)[O-])C(=C2C=C(C(=O)C(=C2)C(=O)O)C)C3=C(C=CC(=C3Cl)S(=O)(=O)[O-])Cl.[Na+].[Na+].[Na+] |
IUPAC Name | trisodium;5-[(3-carboxy-5-methyl-4-oxocyclohexa-2,5-dien-1-ylidene)-(2,6-dichloro-3-sulfonatophenyl)methyl]-3-methyl-2-oxidobenzoate |
Molecular Weight (g/mol) | 605.277 |
PubChem CID | 54736314 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'